missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters

Ten fixed-volume models from 10 to 1000μL

Brand:  VistaLab™ Technologies 1023

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11935782

  • 255.00 € / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • “To deliver” capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Specifications

Specifications

±3.00%
White
1
Precision Pipetter
Nozzle Insert, Calibration Key
Limited Lifetime
2 in.
Not Autoclavable
Aluminum, Stainless Steel
Non-sterile
25 μL
1 Ea.
9 in.
1.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers