missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies MLA™ Brand Precision Pipetters

Ten fixed-volume models from 10 to 1000μL

Brand:  VistaLab™ Technologies 1025

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11955782

  • 160.00 € / Each


Estimated Shipment
08-06-2021
Add to basket

Description

Description

  • “To deliver” capability ensures volume accuracy over a wide range of viscosities
  • Ideal for repetitive delivery of reagents when high precision and accuracy are required
  • Sleeves are color coded for easy volume identification
  • Capacity engraved on bonnet

Order Info

Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.

Specifications

Specifications

±1.00%
1.5 in.
9 in.
Non-sterile
2 in.
Aluminum and Stainless Steel
100 µL
Not Autoclavable
Nozzle Insert, Calibration Key
1 Ea.
Limited Lifetime
Blue
1
Videos
SDS
Documents