missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies MLA™ Brand Precision Pipetters
Ten fixed-volume models from 10 to 1000μL
Brand: VistaLab™ Technologies 1026
Additional Details : Weight : 0.50000kg
Description
- “To deliver” capability ensures volume accuracy over a wide range of viscosities
- Ideal for repetitive delivery of reagents when high precision and accuracy are required
- Sleeves are color coded for easy volume identification
- Capacity engraved on bonnet
Order Info
Additional sizes available. Contact your Fisher Scientific Customer Service Representative for more information.
Specifications
±1.00% | |
Red | |
1 | |
Precision Pipetter | |
Nozzle Insert, Calibration Key | |
Limited Lifetime | |
2 in. |
Not Autoclavable | |
Aluminum, Stainless Steel | |
Non-sterile | |
200 μL | |
1 Ea. | |
9 in. | |
1.5 in. |