Learn More
VistaLab™ Technologies Ovation™ Pipet Nozzle Seals
Nozzle seals and seal accessories for Ovation BioNatural Pipets.
34.01 € - 133.27 €
Product Code | Brand | Volume (Metric) | Quantity | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Quantity | Type | Price | Quantity & Availability | |||
11906042
|
VistaLab™ Technologies
9060-1012 |
0.5 to 20 μL | 24/Pk. | Nozzle Seal |
133.27 €
Pack of 24 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
11916042
|
VistaLab™ Technologies
9060-1013 |
0.5 to 20 μL | 4/Pk. | Nozzle Seal |
34.01 €
Pack of 4 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
11926042
|
VistaLab™ Technologies
9060-2010 |
2.0 to 125, 5.0 to 250 μL | 4/Pk. | Replacement Nozzle |
82.72 €
Pack of 4 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
11956042
|
VistaLab™ Technologies
9060-2013 |
2.0 to 125, 5.0 to 250 μL | 4/Pk. | Nozzle Seal |
34.01 €
Pack of 4 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
11996042
|
VistaLab™ Technologies
9060-3013 |
25 to 1250 μL | 4/Pk. | Nozzle Seal |
34.01 €
Pack of 4 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
11986042
|
VistaLab™ Technologies
9060-3012 |
25 to 1250 μL | 24/Pk. | Nozzle Seal |
133.27 €
Pack of 24 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
Description
- Available in a range of sizes
Specifications
2 in. | |
2 in. | |
5 in. |