Learn More
VistaLab™ Technologies Ovation™ Pipet Nozzle Seals
Nozzle seals and seal accessories for Ovation BioNatural Pipets
Brand:  VistaLab™ Technologies 9060-2013
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 11956042
  • 29.19 € / Pack of 4
Estimated Shipment
11-05-2021
Add to basket
Description
Description
  • Available in a range of sizes
Specifications
Specifications
2 to 125 µL, 5 to 250 µL Ovation™ BioNatural™ Multichannel Pipette
2 in.
Nozzle Seals
2.0 to 125 µL & 5.0 to 250 µL
5 in.
4/Pk.
2 in.