missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Reagent Reservoirs
Unique trough-within-trough design maximizes reagent recovery
Brand: VistaLab™ Technologies 3054-1005
Additional Details : Weight : 0.83700kg
Description
- Polystyrene with trimmed base corners
- Distinct pour-off spouts to empty reservoirs without spills
- Well-marked graduated increments on inside wall minimize reagent waste
- Wide base and sturdy construction provide stability and help to avoid spills
- Molded corner structures allow easy separation of bulk packaged reservoirs
- Available in 25, 50, 100mL and 1.8L volumes; 25mL available with divided configuation for multichannel pipetting of two reagents simultaneously
- 100mL model (Cat. No. 21-385-111) is supplied with lid
- Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
Order Info
Reservoir lid sold separately (Cat. No. 21-385-110).
Specifications
Pyrogen-free | |
Yes | |
13.5 in. | |
50/Cs. | |
25 mL (divided) | |
Sterile |
White | |
12.5 in. | |
Individually Wrapped | |
6 in. | |
Polystyrene | |
Reagent Reservoir |