missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ VITsafe™ Wide-Mouth Wash Bottle

Product Code. 12781178
Click to view available options
Capacity (Metric):
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Vitlab™ VITsafe™ Wide-Mouth Wash Bottle with precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube.

  • Bottles made from PE-LD (or PP for Acetone and MEK), spray tube from PP
  • With coloured VENT-CAP which supports easy visual identification of the media
  • More safety due to the durable safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
  • Practically no leakage or dripping with the VENT-CAP, the design of which prevents almost all static overpressure

Specifications

Diameter (Metric) 76 mm
Height (Metric) 166 mm
Capacity (Metric) 500 mL
Color Red
Includes Xylene Imprint
Material LDPE
Thread Size (Metric) GL 45
Mouth Wide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'