missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Vivacon 2 125kDa

Centrifugal units for fast DNA concentration and buffer exchange of 2 mL samples.

Brand:  Sartorius Stedim Biotech™ VN02H81

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15933040

  • 228.00 € / Pack of 25
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

  • The product of choice for protein fractionation applications.
  • The use of Hydrosart membranes results in high recoveries from dilute samples.
  • Complete and highly reproducible recovery of the concentrate.
  • Reverse spinning step capacity.
  • Concentration of up to 0.5 mL genomic DNA or peptide samples.
  • Low binding material.
  • MWCO printed on device.
  • Selected cut offs available sterile and DNA-free for things like forensic applications.
  • qPCR assay over 55 cycles with a verified limit of detection < 2 pg
Specifications

Specifications

MWCO: 125,000 Daltons
25/Pk.
Polycarbonate Concentrator Body, Polypropylene Filtrate Vessel, Cellulose Acetate Membrane
0.1 to 2.0 mL, 55 μL (Dead Stop Volume)
Centrifugal ultrafiltration device
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers