Learn More
Sartorius™ Vivaspin™ Turbo 4 Centrifugal Concentrators
Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius™ Vivaspin™ Turbo 4 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery for 2 to 4mL sample volumes.
Brand: Sartorius™ VS04T01
Code : NEW
Additional Details : Weight : 0.39480kg
Description
Twin vertical membranes and a sleek internal design for ultra-fast concentration- In a single spin, most 4mL solutions can be concentrated up to 150x in 10–30 minutes
- Large, high-flux PES membranes feature ultra-low protein binding and low extractables
- High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
- Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
- Eliminates the risk of sample filtration to dryness
- Stable, inert polystyrene housing with high chemical compatibility (pH 1–14)
- UV joining technology for a smooth transition between membrane and housing
- Volume graduation for easy monitoring of concentrate volume and 30μl dead-stop volume
- Universal compatibility with nearly all rotors that accommodate 15mL (17mm) conical bottom tubes
Specifications
In either swing bucket or fixed angle rotors accepting 15mL centrifuge tubes | |
10 kDa | |
High flux polyethersulfone membrane range that is recommended for most solutions | |
25Pack | |
4 mL |
PES | |
Concentration of biological samples | |
5000g | |
Centrifugal Concentrator |