missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with Single Certificate

Poulten & Graf GmbH VOLAC FORTUNA™ Bulb Pipette with Single Certificate allow accurate measurements of a volume of solution.

Brand:  Poulten & Graf GmbH R371/WAC/I/5

Code : NEW

Additional Details : Weight : 0.17500kg

 View more versions of this product

Product Code. 15509441

  • 50.10 € / Pack of 5
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Amber graduation
  • With 1 mark
  • Conformity approved
  • With batch certificate
  • With single certificate
Specifications

Specifications

Autoclavable
AS
Non-sterile
Ring marks
0.03 mL
One-mark Bulb Pipette, VOLAC FORTUNA, with single certificate
EN ISO 648
Soda Lime Glass
Amber
5
25 mL
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers