Learn More
Hirschmann™ Volumetric Pipets with Attached Syringe
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann™ Volumetric Pipets with Attached Syringe come with amber graduation.
24.10 € - 82.70 €
Code produit | Marque | Volume (Metric) | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Volume (Metric) | Prix | Quantité et disponibilité | |||||
10544062
|
Hirschmann™
1410160 |
10 mL |
33.40 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10723343
|
Hirschmann™
1410155 |
5 mL |
30.45 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10223771
|
Hirschmann™
1410133 |
1 mL |
24.10 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10643392
|
Hirschmann™
1410165 |
20 mL |
42.30 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10731231
|
Hirschmann™
1410175 |
50 mL |
82.70 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10155480
|
Hirschmann™
1410170 |
25 mL |
47.05 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10577441
|
Hirschmann™
1410164 |
15 mL |
41.05 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
10110621
|
Hirschmann™
1410140 |
2 mL |
26.10 €
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Description
- One mark
- Amber stain graduation
Spécification
Yes | |
Volumetric Pipette Syringe |