missing translation for 'onlineSavingsMsg'
Learn More
Learn More
von Willebrand Factor Mouse, Clone: VWF/1465, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | von Willebrand Factor |
---|---|
Clone | VWF/1465 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961078
|
EnquireBio
7450-MSM5-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951078
|
EnquireBio
7450-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
von Willebrand Factor | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7450 | |
Recombinant fragment of human vWF protein (aa1815-1939) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
250kDa |
VWF/1465 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_000543 | |
VWF | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
HUVEC cells or Tonsil |