Learn More
Emma Safety Footwear Vulcanus Safety Shoes
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
Brand:  Emma Safety Footwear MM53984839
Additional Details : Weight : 1.79700kg
 View more versions of this product
Product Code. 15938701
  • 134.62 € / 1 pair
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Velcro over-foot flap
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Vulcanus
Leather
Black
Full grain Nappa /Hydro-Tec™ Sanitized Silver
39