missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical

Product Code. 31751110
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31751110 500 mL Natural Poly Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31751110 Supplier Ricca Chemical Company Supplier No. R9150000500A
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
CAS Min % 98.0
Melting Point 0°C
Boiling Point 100°C
CAS Max % 100.0
Conductivity max. 0.056
Synonym dihydrogen oxide
Solubility Information Miscible
IUPAC Name water
Molecular Weight (g/mol) 18.02
Formula Weight 18.01
Percent Purity 100%
Grade ACS
Packaging Natural Poly Bottle
Sterility Non-sterile
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water, ASTM Type I
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.