missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Water Bath with Lid
Pan and lid style water bath with two supporting handles. Bochem™ Water Bath with Lid is constructed of 18/10 stainless steel.
186.00 € - 267.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10595221
|
Bochem™
8466 |
5.0 L |
186.00 €
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
10028330
|
Bochem™
8468 |
13.0 L |
267.00 €
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
- Pan and lid.
- 18/10 stainless steel construction.
Specifications
Lid | |
Stainless Steel |