missing translation for 'onlineSavingsMsg'
Learn More

Water, DEPC-Treated, Thermo Scientific Chemicals

Catalog Number 15444539
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

 

It is used as mobile phase solvent in LC-MS analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as mobile phase solvent in LC-MS analysis.

Solubility
Soluble in water. Insoluble in polar liquids.

Notes
Suitable for any general chemical storage area. Protect from freezing. Incompatib
TRUSTED_SUSTAINABILITY
CAS 7732-18-5
Color Colorless
Boiling Point 100°C
Molecular Formula H2O
MDL Number MFCD00011332
Beilstein 2050024
Merck Index 14,10039
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Solubility Information Soluble in water. Insoluble in polar liquids.
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02
Odor Odorless
Density 1.000
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, DEPC-Treated
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'