missing translation for 'onlineSavingsMsg'
Learn More

Water, for molecular biology, DNAse, RNAse and Protease free, Thermo Scientific Chemicals

Catalog Number p-29549
72.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


74.10 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic Bottle
Plastic Drum
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, 100.0 deg.C, CHEBI:15377, 0.1ppm max., Undesignated, 1.0000g/mL, none detected, 16,383, 18.02, Molecular Biology, XLYOFNOQVPJJNP-UHFFFAOYSA-N, oxidane, 1ppb max., 15, 10237, MFCD00011332, 18.015, DNase, RNase, Protease free

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Boiling Point 100.0°C
Linear Formula H2O
Fieser 16383
Merck Index 15, 10237
Formula Weight 18.02
Grade Molecular Biology
Packaging Plastic Bottle
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water, DNase, RNase, Protease free
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'