missing translation for 'onlineSavingsMsg'
Learn More

Water, Nuclease-free, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Catalog Number p-7082768
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
1 L
5 L
Packaging:
Nalgene Bottle
Upright carton
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
MFCD00011332
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Bottle
Density 1
Quantity 10 x 1 mL
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'