Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand: Watson-Marlow 981.0152.000
Additional Details : Weight : 1.00000kg
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Specifications
Pump Tubing | |
1.52 mm | |
0.03 in. | |
323S/MC peristaltic pump | |
0.27 to 19 (four roller pump), 0.2 to 14 (eight roller pump)mL/min. |
1/20 in. | |
PVC | |
0.8 mm | |
1.52mm | |
Yellow/Blue |