missing translation for 'onlineSavingsMsg'
Learn More

Medicom WeePro Labo Sterile Coverall, Type 5/6

Coverall sterilized by gamma irradiation and cleanroom apparel compatible from ISO class 4.

287.00 €

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17829301
View Documents
Medicom
WL-PLMS-02
Medium
287.00 €
Pack of 25
Only null left
Estimated Shipment: 29-08-2025
Add to basket
 
17889301
View Documents
Medicom
WL-PLMS-03
Large
287.00 €
Pack of 25
Only null left
Estimated Shipment: 29-08-2025
Add to basket
 
17839301
View Documents
Medicom
WL-PLMS-04
X-Large
287.00 €
Pack of 25
Only null left
Estimated Shipment: 29-08-2025
Add to basket
 
17899301
View Documents
Medicom
WL-PLMS-05
2X-Large
287.00 €
Pack of 25
Only null left
Estimated Shipment: 29-08-2025
Add to basket
 
Description

Description

  • New material MICROMIUM™, light and very low contaminant
  • Cleanroom apparel compatible from ISO class 4
  • Chemical splash suit, Category III Type 5/6
  • Double packaging fully sealed
  • Apparel sterilized by gamma irradiation
Specifications

Specifications

Cleanroom Coveralls
Microporous Fabric
Single-use
Covered Elastic
Hooded
Zipper
Sterile
None
Unisex
White
Elastic (Stitched-in)
Elastic
3-panel
Bound
No
Class 10-100,000 (ISO 4-8)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.