Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Pipette, Receiver Kit and Filter
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
250.16 € - 350.75 €
Product Code | Brand | Pump Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pump Type | Price | Quantity & Availability | |||||
10700854
|
Welch™
112580-01 |
Hand Held Pipette |
325.42 €
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
10523543
|
Welch™
U1475K-20 |
Spare Receiver Kit |
250.16 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
12759606
|
Welch™
U1475K-21 |
Replacement Hydrophobic Filter |
350.75 €
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
- Bleach resistant construction for extending service life
- Continuous vacuum level monitoring with dial vacuum gauge
- Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station |