missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 4 and 13mm Puradisc Polyethersulfone Syringe Filters

For minimal adsorptive protein loss

Brand:  Cytiva 6782-1302

186.35 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.



Code : 75

Additional Details : Weight : 0.47000kg

 View more versions of this product

Product Code. 11304674

  • 255.00 € / Pack of 100
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Description

Description

Suitable for filtering serum, plasma and tissue culture solutions—as well as other protein-containing solutions where minimal adsorptive protein loss is required.

  • Polypropylene housing
  • Autoclavable
  • Female Luer lock on inlet side; male slip Luer on outlet side

4mm dia. Filters

  • For samples up to 2mL
  • Filter area: 0.2cm2 Hold-up volume (with air purge) <7.5μL
13mm dia. Filters
  • For samples up to 10mL
  • Filter area: 1.3cm2
  • Hold-up volume (after air purge) <25μL

Specifications

Specifications

Autoclavable
Polysulfone
Non-sterile
≤25 μL
Male Luer Slip
Polypropylene
≤10 mL
Passes USP Class VI requirements for biosafety.
0.2 μm
For filtering serum, plasma and tissue culture solutions – as well as other protein-containing solutions where minimal adsorptive protein loss is required.
Female Luer Lok
100/Pk.
Syringe Filter
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers