Learn More
Cytiva Whatman™ 850-DS 8-Channel Filter Plate
Use with Agilent™ 850-DS Dissolution Sampling Station for automated sample preparation in dissolution testing
Brand:  Cytiva 7707-3900
Additional Details : Weight : 1.70000kg
Product Code. 16304566
  • 910.00 € / Pack of 50
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
Description
Description
  • Exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station
  • Simplifies filter replacement between timepoints
  • Reliable alignment of the liquid path increases productivity by reducing the risk of jamming, and reducing leaks that may occur with other dissolution sample preparation systems
  • Save time and eliminate errors associated with manual sampling
  • Automated processing of up to 8 samples simultaneously
  • Readily available in a wide range of pore sizes and materials
Specifications
Specifications
Agilent 850-DS Dissolution Sampling Station
Automated Sample Preparation in Dissolution Testing
8 Channel
Glass Microfiber
50/Pk.