missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Acid/Alkali Test Papers

Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.

Brand:  Cytiva 2600-602

Code : 69

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 11804473

  • 103.00 € / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Assumes a color specific to sample pH upon contact with a sample. Whatman™ Acid/alkali Test Papers are impregnated with a mixture of pH indicators.

  • Litmus Blue turns red in acid
  • Litmus Red turns blue in base, at pH 5 to 8
  • Congo Red ph range: 6.8 to 8.3

Specifications

Specifications

Litmus Red
Strip
pH Test Paper
10 mm
5 to 8
10 x 10 Books of 20 Strips
60 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers