missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Autovial™ 5mL Syringeless Filters

Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.

Brand:  Cytiva AV115NPUNYL

Code : 69

Additional Details : Weight : 0.95000kg

 View more versions of this product

Product Code. 11373185

  • 178.00 € / Pack of 50
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

 

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Remove particulates from samples up to 5mL in volume
  • Filtration area: 1.7cm2

 

Glass Microfiber (GMF)

  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
  • Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
  • Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
  • Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples

 

Specifications

Specifications

Nylon
Non-sterile
Syringeless Filter
13 mm
0.45 μm
50/Pk.
≤5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers