Learn More
Cytiva Whatman™ Autovial™ 5mL Syringeless Filters
Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.
Brand: Cytiva AV115NPUORG
191.28 EUR valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Code : 69
Additional Details : Weight : 0.45000kg
Description
Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Remove particulates from samples up to 5mL in volume
- Filtration area: 1.7cm2
Glass Microfiber (GMF)
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Nylon (NYL)
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- Polytetrafluoroethylene (PTFE)
- Hydrophobic; for samples >50% organic solvents
- Polyvinylidene Fluoride (PVDF)
- Low protein binding; for aqueous samples
Specifications
PTFE | |
Non-sterile | |
Syringeless Filter | |
13 mm |
0.45 μm | |
50/Pk. | |
≤5 mL |