Learn More
Cytiva Whatman™ Benchkote Plus Surface Protector Sheets
Prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. Whatman™ Benchkote Plus bench liner can also help with the disposal of hazardous materials.
Brand: Cytiva 2301-6150
115.37 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : 03
Additional Details : Weight : 2.38950kg
Description
Prevent many common sources of laboratory contamination, such as those involving hazardous materials on shared workspace benches. Whatman™ Benchkote Plus bench liner can also help with the disposal of hazardous materials.
- Strong: Benchkote Plus bench liner is strong and tear resistant when wet or dry
- Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
- Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
- Disposable: Benchkote Plus bench paper can be incinerated after use
- Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
Specifications
Surface Protector | |
750 mL/m2 | |
500 mm | |
600 mm | |
Suitable for saturation with disinfectant |
Polyethylene | |
19 in. | |
23 in. | |
500 x 600mm | |
50 Sheets |