Learn More
QIAGEN QIAcard Bloodstain
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
Brand: QIAGEN WB100014
Code : A0
Additional Details : Weight : 0.42000kg
Description
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
- Non-Indicating and Non-FTA treated card designed for the collection, short-term handling of specimens.
- Unlike FTA Cards, the Bloodstain Cards are not coated with a protective chemical cocktail and DNA is as risk of degradation and damage.
- Convenient transport of blood and bodily fluids with protective cover included.
Specifications
Card | |
Collection and transport of blood and body fluids | |
100 Pk. | |
125μL per sample area |