Learn More
Cytiva Whatman™ Drying Rack for 903 Protein Saver Cards
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
36.29 € - 102.00 €
Specifications
Product Type | Drying Rack |
---|---|
For Use With (Equipment) | 903 Protein Saver Card |
Quantity | 10 Pack |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
11909134
|
Cytiva
10539521 |
Hook-and-Loop Mounting Sticker |
102.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11359394
|
Cytiva
10537173 |
Mounting Sticker not included |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Air-dries blood specimens in a suspended horizontal position. Whatman™ Drying Rack for 903 Protein Saver Card is easily assembled and can be affixed to a wall or counter top with optional hook-and-loop stickers.
- For use with neonatal human diagnostic testing cards
- CE Marked
- Accommodate multiple collection forms at one time
Specifications
Drying Rack | |
903 Protein Saver Card | |
10 Pack |