missing translation for 'onlineSavingsMsg'
Learn More

Qiagen FTA™ Elute Cards

Provides room temperature DNA storage combined with simplified purification of DNA for multiple amplifications from a single sample. Qiagen FTA™ Elute Cards contain a chaotropic salt keeping proteins tightly bound while DNA is eluted from the matrix.

Brand:  Qiagen WB120401

Code : 69

Additional Details : CAS Number : 593-84-0 Weight : 0.06300kg

 View more versions of this product

Product Code. 11938117

Quantity Price
1 137.00 € / Pack of 25
10 115.12 € / Pack of 25
Estimated Shipment: 07-07-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

 

Non-Indicating FTA Elute Micro Cards enable you to collect, process, transport, and store colored biological samples, such as blood samples.

  • Improves process efficiency by enabling direct amplification for STR analysis.
  • Ideal for colored samples, like blood.
  • Immediate stabilization and protection of nucleic acids due to FTA technology.
  • Different card options to support manual or automated processing for fast and efficient sampling.
  • Long term archiving of samples at room temperature due to FTA card technology.
  • Hemoglobin is bound to matrix, that acts as PCR inhibitor
  • Yields DNA in solution
  • Sample processing time to isolate DNA: 15 to 30 minutes
  • Sample processing: hot water elution procedure
  • Sample volume: 12 to 40μL per collection area
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection
Specifications

Specifications

Multiplex PCR.Sequencing after PCR amplification.SNP analysis.STR analysis.HLA typing.Whole genome amplification.Quantitative PCR.Biobanking.Pharmacogenomics.Genotyping.Genetic identification.Transgenic detection.
Blood
4
25 Pieces
Room Temperature
40 μl
15 min to 30 min
Collect, process, transport and store colored biological samples, such as blood samples with Whatman Non-Indicating FTA Elute Micro Cards.
FTA Elute Micro Card, 4 sample areas per card
Chaotropic
15
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .

SDS
Documents
Certificates