Learn More
QIAGEN Resealable Multi-Barrier Pouches
For contamination-free collection, storage and transport of DNA samples.
Brand: QIAGEN WB100024
Code : 50
Additional Details : Weight : 0.49000kg
Description
- Resealable Multi-Barrier Pouches for transport or storage of QIAcard FTA CloneSaver.
- Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
- The outer paper surface is suitable for labeling or writing.
- The Multi-Barrier Pouch has a resealable closure.
- To optimize sample integrity FTA Cards are stored in a Multi-Barrier Pouch with a desiccant packet.
Specifications
CloneSaver™ Card | |
Collection, storage, processing and shipping FTA™ and FTA™ elute cards | |
Multi-barrier Pouch | |
12 x15 cm |