Learn More
Cytiva Whatman™ Funnels for Microbiological Filtration System
Autoclavable plastic funnel for the MBS I system.
Brand: Cytiva 10445866
Code : 52
Additional Details : Weight : 0.42300kg
Description
- Simple to use.
- Safe sealing mechanism.
- Shorter preparation time.
- High reproducibility.
- Funnels can be autoclaved up to 50 times.
- Large funnel capacity for foaming liquids.
- Easy to validate.
- Risk of cross-contamination is minimized.
Specifications
PP Funnel | |
Microbiological filtration system (MBS I) | |
350 mL |