Learn More
Cytiva Whatman™ Glass Microfiber Filter Paper Grade 6
Captures tiny particles without becoming weak. Whatman™ Glass Microfiber Filter Paper Grade 6 may also be used for determination of filterable substances and residue upon ignition or analysis of aggressive media.
77.60 € - 920.00 €
Specifications
Material | Borosilicate Glass |
---|---|
Particle Retention | <1 μm |
Filtration Speed | 40 s/100mL/in2 |
Flow Rate | Slow |
Grade | GF 6 |
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Format | Price | Quantity & Availability | ||||
11385254
|
Cytiva
10370002 |
50 mm | Disc |
122.00 €
Pack of 200 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
11305264
|
Cytiva
10370004 |
70 mm | Disc |
141.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11395254
|
Cytiva
10370003 |
55 mm | Disc |
111.00 €
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11315264
|
Cytiva
10370005 |
90 mm | Disc |
196.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11325264
|
Cytiva
10370006 |
110 mm | Disc |
318.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11335264
|
Cytiva
10370007 |
125 mm | Disc |
335.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11345264
|
Cytiva
10370008 |
150 mm | Disc |
459.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11355264
|
Cytiva
10370010 |
185 mm | Disc |
515.00 €
Pack of 100 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
11375264
|
Cytiva
10370012 |
240 mm | Disc |
675.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11365264
|
Cytiva
10370011 |
200 mm | Disc |
595.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11305274
|
Cytiva
10370020 |
100 mm | Disc |
311.00 €
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11395264
|
Cytiva
10370019 |
47 mm | Disc |
109.00 €
Pack of 200 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
11385264
|
Cytiva
10370018 |
25 mm | Disc |
77.60 €
Pack of 200 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
11315274
|
Cytiva
10370050 |
620 mm | Sheet |
920.00 €
Pack of 100 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||||
Description
Suitable for water and wastewater analysis, with extremely fine particle retention and added strength.- Sub micron filter retention efficiency to capture extremely small particles
- Inorganic binder for increased strength
- Temperature-resistant up to 500°C
- Can be used as a pre-filter for dense samples
Specifications
Borosilicate Glass | |
40 s/100mL/in2 | |
GF 6 |
<1 μm | |
Slow | |
350 μm |