missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman Grade GF/C Glass Microfiber Filters, Binder Free, Ready to Use

Grade GF/C glass filters offer fine particle retention, good flow rate and loading capacity, and support total suspended solids analysis.

Brand:  Cytiva 2822-047

Additional Details : Weight : 0.01800kg


 View more versions of this product

Product Code. 15853704

  • 98.15 € / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Good flow rate and loading capacity
  • Fine nominal particle retention of 1.2μm
  • Binder free
  • Applications include total suspended solids analysis, industrial waste analysis and potable water
  • GF/C RTU pre-treats traditional GF/C filter with washing, drying, cooling (in a desiccator), and weighing steps aligned to EN 872
  • Certified weight loss of less than 0.017mg/cm2 after preparation
  • Each RTU filter is placed in an individually barcoded aluminum pan that notes the filter weight
Specifications

Specifications

6.7s/100mL/in2
Borosilicate Glass
1.2 μm
Grade GF/C
550°C
Filter, pre-rinsed and dried
4.7 cm
GE’s ready-to-use Whatman filter paper Grade GF/C glass filters offer a good flow rate and loading capacity that support total suspended solids analysis with a fine nominal particle retention of 1.2 μm.
1.2 μm
Widely used for cell harvesting, liquid scintillation counting and binding assays where more loading capacity is required.Highly suitable for use in applications involving air filtration and for gravimetric analysis of volatile materials where ignition is
100 Pieces
260 μm
105 mL/min
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FILTER GLASS CIRCLE

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glass, oxide, chemicals
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers