Learn More
Cytiva Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
105.00 € - 299.00 €
Specifications
Description | GE’s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis. |
---|---|
Material | Cellulose |
For Use With (Application) | Air and waste gas analysis |
For Use With (Equipment) | Soxhlet extractor |
Max. Temperature (Metric) | 120°C |
Product Code | Brand | Length (Metric) | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inside | Price | Quantity & Availability | ||||
11303225
|
Cytiva
2800-226 |
65 mm | 22 mm |
113.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
10536731
|
Cytiva
2800-228 |
80 mm | 22 mm |
112.00 €
Pack of 25 |
In Stock
Log in to see stock. |
||||
11330704
|
Cytiva
2800-266 |
60 mm | 26 mm |
134.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11310714
|
Cytiva
2800-338 |
80 mm | 33 mm |
140.00 €
Pack of 25 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
11350714
|
Cytiva
2800-432 |
123 mm | 43 mm |
191.00 €
Pack of 25 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
11310704
|
Cytiva
2800-250 |
100 mm | 25 mm |
120.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11390694
|
Cytiva
2800-199 |
90 mm | 19 mm |
111.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11320714
|
Cytiva
2800-339 |
94 mm | 33 mm |
133.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11360714
|
Cytiva
2800-608 |
180 mm | 60 mm |
299.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11313225
|
Cytiva
2800-259 |
90 mm | 25 mm |
120.00 €
Pack of 25 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11370704
|
Cytiva
2800-300 |
100 mm | 30 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11380704
|
Cytiva
2800-308 |
80 mm | 30 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11300714
|
Cytiva
2800-331 |
118 mm | 33 mm |
172.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11370694
|
Cytiva
2800-105 |
50 mm | 10 mm |
113.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11320704
|
Cytiva
2800-258 |
80 mm | 25 mm |
120.00 €
Pack of 25 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
11350704
|
Cytiva
2800-282 |
120 mm | 28 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11360704
|
Cytiva
2800-288 |
80 mm | 28 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11330714
|
Cytiva
2800-373 |
130 mm | 37 mm |
178.00 €
Pack of 25 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
11340714
|
Cytiva
2800-412 |
123 mm | 41 mm |
191.00 €
Pack of 25 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
11340704
|
Cytiva
2800-280 |
100 mm | 28 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11323225
|
Cytiva
2800-260 |
100 mm | 26 mm |
120.00 €
Pack of 25 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
11390704
|
Cytiva
2800-330 |
100 mm | 33 mm |
122.00 €
Pack of 25 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
10419111
|
Cytiva
2800-185 |
55 mm | 18 mm |
105.00 €
Pack of 25 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
||||
Description
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
- Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
- Suitable for temperatures up to 120°C (248°F)
- Excellent mechanical strength and retention
- Wall thickness of standard thimbles is 1mm to support application need
Specifications
GE’s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis. | |
Air and waste gas analysis | |
120°C |
Cellulose | |
Soxhlet extractor | |
Extraction Thimble |