Learn More
Cytiva Whatman™ Membrane-Butler
Optimal handling for all MicroPlus and ME membrane filters
Brand: Cytiva 10477100
Code : 50
Additional Details : Weight : 3.32000kg
Description
- High reliability
- Minimizes cross contamination risks
- Membrane dispenses rapidly
- Ideal for sterile benches
- Compact dimensions
Specifications
Manual membrane butler | |
Dispensing filtration membranes | |
Sterile | |
1/Pk. |