missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Qualitative Filter Paper: Grade 1 Sheets
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Sheets are widely used standard grade filter paper for routine applications with medium retention and flow rates.
75.75 € - 935.00 €
Specifications
Format | Sheet |
---|---|
Material | Cellulose |
Pore Size | 11 μm |
Filtration Speed | 150 sec. |
Flow Rate | Medium |
Product Code | Brand | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
11392815
|
Cytiva
1001-813 |
26 x 31 mm (1 x 1.23 in.) | 1000Pack |
76.72 €
Pack of 1000 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
11302825
|
Cytiva
1001-824 |
75 x 100 mm (3 x 4 in.) | 500 Pack |
75.75 €
Pack of 500 |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
11312825
|
Cytiva
1001-917 |
460 x 570 mm (18.11 x 22.45 in.) | 100 Pack |
206.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
11322825
|
Cytiva
1001-918 |
460 x 570 mm (18.11 x 22.45 in.) | 500 Pack |
935.00 €
Pack of 500 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
11310404
|
Cytiva
1001-929 |
600 x 600 mm (23.62 x 23.62 in.) | 100 Pack |
205.00 €
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
11332825
|
Cytiva
1001-931 |
580 x 680 mm (22 x 26 in.) | 100 Pack |
262.00 €
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
11300404
|
Cytiva
1001-932 |
580 x 680 mm (22 x 26 in.) | 500 Pack |
830.00 €
Pack of 500 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
Description
Covers a wide range of laboratory applications and is frequently used for clarifying liquids. Whatman™ Qualitative Filter Paper: Grade 1 Sheets are widely used standard grade filter paper for routine applications with medium retention and flow rates.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids.
- Particle Retention: >11μm
- Porosity: medium
- Filtration speed: ASTM™, 40 sec.; Herzberg, 150 sec.
- Surface: smooth
- Separates precipitates such as lead sulfate, calcium oxalate (hot) and calcium carbonate
- Used for soil analysis and seed testing procedures in the agricultural industry
- Used to separate solid foodstuffs from associated liquid or extract liquid in the food industry
- Atmospheric dust is collected from airflow and the stain-intensity measured
- Impregnated with a chromogenic reagent and color formation quantified by optical reflectance for gas detection
Specifications
Sheet | |
11 μm | |
Medium | |
Grade 1 | |
180 μm |
Cellulose | |
150 sec. | |
A widely used standard grade filter paper for routine applications with medium retention and flow rate. It covers a wide range of laboratory applications and is frequently used for clarifying liquids. | |
Medium |