missing translation for 'onlineSavingsMsg'
Learn More

Qiagen Uni-Core Punch

Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device

Brand:  Qiagen WB100039

Code : A0

Additional Details : Weight : 0.90000kg

 View more versions of this product

Product Code. 11937907

  • 59.44 € / Pack of 4

Estimated Shipment
29-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Disposable punch for use with FTA cards, FTA Elute Cards, and EasiCollect device

  • Punch with polished steel tip that is case-hardened and can be sterilized
  • Stainless steel cutting edges enable manual coring, retrieval, and controlled ejection of FTA punch
  • Housed in plastic handle
  • Each tip provides up to 500 punches before disposal
  • Available in multiple diameters varying from 0.35 to 12mm
  • Manual punching with low sample volumes punching 1 to 50 samples per day
  • Minimum contamination and carryover between samples when cleaning procedure is followed

Specifications

Specifications

Whatman™ FTA™ cards, FTA™ elute cards and EasiCollect device
3.0mm
Accessory, FTA Card
0.11 in.
Two cutting mats
4 Pack
Videos
SDS
Documents