Learn More
DWK Life Sciences Wheaton™ Capcluster Mat
Used to seal 96-well rack of Microtubes
Brand: DWK Life Sciences W240860
Code : A0
Additional Details : Weight : 0.18140kg
Description
- Thermo Plastic Elastomer (TPE) made
- Multi-pierceable closure
- Resistant to DMSO, other chemical
- Rnase, DNase free
Specifications
TPE | |
10 | |
Capcluster Mat |
96 | |
-112°F to 176°F | |
-80°C to +80°C |