Learn More
DWK Life Sciences Wheaton™ Glass Base for Stainless-Steel Support
For 25mm vacuum filtration assembly
Brand: DWK Life Sciences 419333
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Designed to handle small volumes of liquids for analysis of particulate or microbiological contamination
Specifications
Support Base | |
25mm Vacuum Filtration Assembly | |
Glass |