missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Serum Bottles and Vials
436.05 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Use promo code "23261" to get your promotional price.
Click to view available options
Capacity:
0.1 oz., 2 mL
0.17 oz., 5 mL
0.17 oz., 5mL
0.34 oz., 10 mL
0.7 oz., 20 mL
1.01 oz., 30 mL
1.7 oz., 50 mL
2.03 oz., 60 mL
3.38 oz., 100 mL
4.23 oz., 125 mL
Color:
Amber
Clear
Height (English):
1.41 in.
1.85 in.
2.12 in.
2.28 in.
2.44 in.
2.48 in.
2.87 in.
3.58 in.
3.74 in.
4.21 in.
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Compliance
Meet requirements for Class A of the United States Pharmacopeia for Type I glass.
Specifications
Capacity | 0.17 oz., 5 mL |
Diameter (Metric) | 23 mm |
Height (English) | 1.85 in. |
Height (Metric) | 47 mm |
Capacity (Metric) | 5 mL |
Capacity (English) | 0.17 oz. |
Color | Amber |
Material | Borosilicate Glass |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'