Learn More
DWK Life Sciences Wheaton™ Shorty Vials™ Glass Sample Vials in Lab File™: with Caps
Come in convenient Lab File partitioned trays
Brand:  DWK Life Sciences W225292
Additional Details : Weight : 1.63290kg
 View more versions of this product
Product Code. 12424991
  • 191.00 € / Pack of 200
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
Offering a variety of uses ranging from sample storage to freeze-drying.
  • Clear vials made from borosilicate glass
  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Shorty Vials feature a low profile for greater stability and less dead volume
  • Supplied with attached caps to help maintain cleanliness or without caps (replacement caps can be purchased separately)
  • Vials and caps are autoclavable
  • Selected items fit M-T Vial File™—polystyrene trays with partitions that provide an easy way to inventory samples or to store empty vials
Clear Vials
  • Made from low extractable borosilicate glass to provide superior chemical resistance
  • Conform to USP Type I and ASTM E 438 Type I, Class A requirements
Cap Liners
  • 14B rubber-lined black phenolic screw cap for aqueous samples
  • PTFE/14B rubber-lined black phenolic screw cap for organic samples
Specifications
Specifications
14B Rubber
17 mm
4 mL
Screw
Phenolic
Screw Cap
ASTM E438, USP Type I, Type I, Class A
Clear
Borosilicate Glass (Low Extractable)
For uses ranging from sample storage to freeze drying
1.08 dr.
Autoclavable
41 mm
Vial Rack