Learn More
Buerkle™ White Dosing Spoon
Volumetric and dosing spoons can be used to precisely measure small amounts of powder, granules, and liquids.
28.30 € - 156.00 €
Specifications
Product Type | Sampler |
---|---|
Material | Polystyrene |
Description | Dosing spoons with stripper edge for exact measuring |
Disposable | Yes |
Color | White |
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Price | Quantity & Availability | ||||
16525220
|
Buerkle™
5378-4004 |
0.5 mL | Non-sterile |
28.30 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16535220
|
Buerkle™
5378-4005 |
0.5 mL | Sterile |
55.30 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16545220
|
Buerkle™
5378-4006 |
1 mL | Non-sterile |
31.15 €
Pack of 100 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
||||
16555220
|
Buerkle™
5378-4007 |
1 mL | Sterile |
58.35 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16565220
|
Buerkle™
5378-4008 |
2.5 mL | Non-sterile |
34.45 €
Pack of 100 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
16575220
|
Buerkle™
5378-4009 |
2.5 mL | Sterile |
61.60 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16585220
|
Buerkle™
5378-4010 |
5 mL | Non-sterile |
37.75 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16595220
|
Buerkle™
5378-4011 |
5 mL | Sterile |
66.45 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16505230
|
Buerkle™
5378-4012 |
10 mL | Non-sterile |
40.80 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16515230
|
Buerkle™
5378-4013 |
10 mL | Sterile |
69.95 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16525230
|
Buerkle™
5378-4014 |
15 mL | Non-sterile |
43.50 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16535230
|
Buerkle™
5378-4015 |
15 mL | Sterile |
73.20 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16545230
|
Buerkle™
5378-4016 |
25 mL | Non-sterile |
49.90 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16555230
|
Buerkle™
5378-4017 |
25 mL | Sterile |
84.90 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16565230
|
Buerkle™
5378-4018 |
50 mL | Non-sterile |
71.65 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
16575230
|
Buerkle™
5378-4019 |
50 mL | Sterile |
156.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- The content volume is engraved on the handle
- The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, allowing for good emptying of small remaining amounts
- The flat base makes it possible to set the volumetric spoon down without spilling the content
- The disposable dosing spoon is suitable for use as a sample spoon for taking small sample quantities
- Since the plastic volumetric spoon was developed for single use, there is no cleaning and the possibility of cross-contamination and impurities is excluded
- The dosing spoons and volumetric spoons are suitable for precise measuring for chemical and pharmaceutical applications, cosmetics, animal feed, and the food and beverage industry
- Sterile versions are individually packaged
- With stripper edge
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
Specifications
Sampler | |
Dosing spoons with stripper edge for exact measuring | |
White | |
Especially for The Food, Animal Feed and Pharmaceutical Industries |
Polystyrene | |
Yes | |
Food Contact, Reach, RoHS | |
100/Pk. |