Learn More
Azlon™ Wide Mouth Vented Wash Bottles
Manufactured from LPDE for strength and flexibilty. Azlon™ Wide Mouth Vented Wash Bottles feature color coded lids for easy identification.
Brand:  Azlon WGW445VT
Code : NEW
Additional Details : Weight : 0.07700kg
 View more versions of this product
Product Code. 15397047
  • 28.05 € / Pack of 5
Estimated Shipment
05-02-2019
Add to basket
Description
Description
  • Square shoulder wash bottles
  • Wide neck for easy spill-free filing; patented DripLok™  vapor venting valve prevents solvent drips
  • Color-coded one-piece polypropylene closures maintain shape in use
  • Precision jet-tip aperture for fine control
  • Translucent bottles without print
Specifications
Specifications
1000mL
LDPE
Wide
Polypropylene
Red
Screw cap with curved aperture
Videos
Documentation
Technical Documents