missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilm s Tumor 1 Mouse, Clone: rWT1/857, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7490-MSM6-P1
Additional Details : Weight : 0.01000kg
Specifications
Wilm s Tumor 1 | |
Cytoplasmic | |
rWT1/857 | |
Unconjugated | |
NP_001185480 | |
Mouse | |
47-55kDa | |
Protein A/G Purified | |
RUO | |
7490 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
WT1 | |
Recombinant full-length human WT1 protein | |
K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |