missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes
Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes fit tightly in any spinner turbine and have minimal NMR background.
Brand: Bel-Art™ 66000-0083
Code : NEW
Additional Details : Weight : 0.02700kg
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Description
- High quality 5mm O.D. economy sample tube
- Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
- 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
- Fit tightly in any spinner turbine
- Minimal NMR background
- Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
- Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
- Manufactured in an ISO 9001:2008 USA facility
Specifications
Borosilicate Glass | |
4.947±0.019 mm | |
Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only. | |
7 in. | |
5/Pk. |
178 mm | |
100MHz | |
Tube and disposable cap | |
0.43 mm | |
Wilmad™ WG-5MM-ECONOMY-7-5 |