Learn More
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
23.57 € - 248.88 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15328608
|
Bel-Art™
66000-0038 |
Omni-Fit valve |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15318608
|
Bel-Art™
66000-0037 |
Omni-Fit Septum |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15398858
|
Bel-Art™
66306-0571 |
300mghz Omni-Fit NMR sample tube |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
- Omni-Fit Sample tubes (507-OF-7) sold separately
- Omni-Fit valve (OFV) is a complete accessory set
- Additional septum (OF-30) are available in packages of 6
Specifications
5.0 mm |