Learn More
Wilmad-LabGlass™ SP Scienceware™ Rotor and Caps for Bruker MAS Probe
Provide solution for analysis of solid samples
Brand: Wilmad-LabGlass™ 66809-0581
Code : NEW
Additional Details : Weight : 0.04310kg
Description
- Manufactured from high-purity Zirconia
- Caps fitted with O-rings for improved sealing
- MAS Probe and caps compatible for most solid state NMR spectrometers
- Thoroughly inspected before and after precision-machining process ensures no material irregularities
- Spin testing performed to highest specified spinning speed, assuring performance without overspinning the rotor
Specifications
Vespel | |
Bottom plug | |
520-1A |
For Bruker MAS 3.2mm Probe | |
Solid state NMR spectrometers |