missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) wwPTFE Membrane Disc Filters

Membrane with chemical compatibility for aqueous solutions and aggressive solvents.

Brand:  Cytiva (formerly Pall Lab) 60551

574.85 EUR valid until 2024-12-31
Use promo code "21931" to get your promotional price.



Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 17154521

  • 685.00 € / Pack of 75
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Maximum versatility
  • Filters aqueous solutions or aggressive chemicals
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications

Specifications

wwPTFE
Non-sterile
90 mm
0.45 μm
280 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers