Learn More
XRCC2 Mouse, anti-Human, CoraLite 555, Clone: 3D9A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL555-66652-100UL
Description
The XRCC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human XRCC2. This antibody recognizes human, mouse antigen. The XRCC2 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
| XRCC2 | |
| 3D9A1 | |
| CoraLite 555 | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 155 kDa | |
| IgG2b |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| DKFZp781P0919, DNA repair protein XRCC2, XRCC2 | |
| 100 μL | |
| 7516 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |